Learn More
Cygnus Technologies HUMAN TOTAL IGG ELISA KIT
Supplier:  Cygnus Technologies F160
Encompass
This kit is intended for use in quantifying very low concentrations of total human immunoglobulin G. The limit of detection is less than 0.1ng/mL.
Catalog No. NC9709129