Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total MMP-7 Quantikine ELISA Kit

Catalog No. DMP700
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DMP700 1 Plate
SMP700 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DMP700 Supplier R&D Systems™ Supplier No. DMP700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Total MMP-7 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Total MMP-7 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-7 in cell culture supernates (25 ul), serum (25 ul), heparin plasma (25 ul), saliva (25 ul), urine (50 ul).

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.094 ng/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human MMP-7
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4316
Gene Symbol MMP7
Quantity 1 Plate
Sample Volume Cell Culture Supernates (25uL),Serum (25uL),Heparin Plasma (25uL),Saliva (25uL),Urine (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase
Target MMP-7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.