Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Total Serpin E1/PAI-1 DuoSet ELISA

Catalog No. DY938705
Change view
Click to view available options
Quantity:
1 Kit (5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY938705 1 Kit (5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY938705 Supplier R&D Systems™ Supplier No. DY938705
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Human Total Serpin E1/PAI-1 DuoSet ELISA contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant human free and complexed Serpin E1 (Total Serpin E1).

The Human Total Serpin E1/PAI-1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Serpin E1/PAI-1.

Specifications

Assay Range 7.81 - 500pg/mL
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 5054
Quantity 1 Kit (5 Plates)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.