Learn More
BIOHIPPO Human TPA Microsample
SDP
Catalog No. 5000314037
Encompass
Human TPA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TPA in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TPA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TPA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TPA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14037 Supplier BIOHIPPO Supplier No. BHE15212049
Human TPA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TPA in Serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TPA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TPA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TPA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color