Learn More
ACROBiosystems ACROBiosystems Human TPBG / 5T4 Protein, Fc Tag
Catalog No. 502106895
Encompass_Preferred
Recombinant Protein;100UG;Human TPBG, Fc Tag (TPG-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession # Q13641-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 61.8 kDa.
Catalog No. 50-210-6895 Supplier ACROBiosystems Supplier No. TPGH5253100UG
Recombinant Protein;100UG;Human TPBG, Fc Tag (TPG-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession # Q13641-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 61.8 kDa.