Learn More
Sino Biological Recombinant Human 5T4/TPBG Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 19845H08H1
Encompass_Preferred
A DNA sequence encoding the human TPBG (NP_006661.1) (Asp60-Asp345) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-2485