Learn More
Sino Biological Human TPH1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10473H02H500
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Tryptophan Hydroxylase 1 / TPH1 (rh Tryptophan Hydroxylase 1 / TPH1; Catalog#11931-H07E; P17752-1; Ile2-Ile444).
Catalog No. 50-237-4398