Learn More
Sino Biological Anti-Thyroid peroxidase/TPO Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 205446T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Thyroid peroxidase / TPO fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3876