Learn More
Sino Biological Anti-TPPP3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14360T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TPPP3 (rh TPPP3; Catalog#14360-H07E; Q9BW30; Met1-Lys176). TPPP3 specific IgG was purified by Human TPPP3 affinity chromatography.
Catalog No. 50-240-8422