Learn More
BIOHIPPO Human TPS Microsample
SDP
Catalog No. 5000314531
Encompass
Human TPS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TPS in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 065 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TPS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TPS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TPS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14531 Supplier BIOHIPPO Supplier No. BHE15212179
Human TPS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TPS in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 065 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TPS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TPS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TPS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color