Learn More
Sino Biological Human TPSB2 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10505MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TPSB2 / Tryptase Beta 2 (rh TPSB2 / Tryptase Beta 2; NP_077078.5; Met 1-Pro 275). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2802