Learn More
BIOHIPPO Human TRA2b Microsample
SDP
Catalog No. 5000314118
Encompass
Human TRA2b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TRA2b in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 055 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TRA2b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TRA2b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TRA2b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14118 Supplier BIOHIPPO Supplier No. BHE15210791
Human TRA2b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TRA2b in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 055 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TRA2b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TRA2b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TRA2b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color