Learn More
Sciencell Research Laboratories Human Trabecular Meshwork Cells (HTMC), Cryopreserved, Passage one, >5 x 10^5 cells per vial, Characterized for α-smooth muscle actin, Expansion guarantee 15 population doublings
Catalog No. NC3442669
Encompass
Human Trabecular Meshwork Cells (HTMC), Cryopreserved, Passage one, >5 x 10^5 cells per vial, Characterized for α-smooth muscle actin, Expansion guarantee 15 population doublings
Catalog No. NC3442669 Supplier Sciencell Research Laboratories Supplier No. 6590KIT
Human Trabecular Meshwork Cells (HTMC), Cryopreserved, Passage one, >5 x 10^5 cells per vial, Characterized for α-smooth muscle actin, Expansion guarantee 15 population doublings