Learn More
Abcam Human TRAF2 ELISA kit, 1 x 96 Tests
Catalog No. 502657387
Encompass_Preferred
Human TRAF2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human TRAF2 with a sensitivity of 1.5 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-7387 Supplier Abcam Supplier No. AB2268931X96TESTS
Human TRAF2 ELISA kit is a single-wash 90-min Simplestep used to quantify Human TRAF2 with a sensitivity of 1.5 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement