Learn More
BIOHIPPO Human TRAF2 Microsample
SDP
Catalog No. 5000315817
Encompass
Human TRAF2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TRAF2 in serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 25 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TRAF2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TRAF2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TRAF2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15817 Supplier BIOHIPPO Supplier No. BHE15210686
Human TRAF2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TRAF2 in serum plasma and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 25 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human TRAF2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human TRAF2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human TRAF2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color