Learn More
Sino Biological Human IL4R / CD124 Protein (His Tag) 500ug
Supplier:  Sino Biological 10402H08H500
Encompass_Preferred
A DNA sequence encoding the human IL4R (NP_000409.1) precursor (Met 1-His 232) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-222-3532