Learn More
ACROBiosystems ACROBiosystems Human TRAIL R1 / DR4 / TNFRSF10A Protein, Fc Tag
Supplier:  ACROBiosystems TR1H52541MG
Encompass_Preferred
Recombinant Protein;1MG;Human TRAIL R1, Fc Tag (TR1-H5254) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Asn 239 (Accession # NP_003835).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.0 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TRAIL R1, Fc Tag (Cat. No. TR1-H5254) at 5 μg/mL (100 μL/well) can bind Human TRAIL, Mouse IgG2a Fc Tag (Cat. No. TRL-H5259) with a linear range of 4-16 ng/mL (QC tested).
Catalog No. 50-210-6756