Learn More
Biomatik Corporation Human Transcription Factor 7 Like Protein 2 (TCF7L2) ELISA Kit
Supplier:  Biomatik Corporation EKU10243
Encompass_Preferred
Human Transcription Factor 7 Like Protein 2 (TCF7L2) ELISA Kit for detecting TCF7L2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NQB0. Gene ID: 6934. Target Synonyms: TCF4; HMG box transcription factor 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5762