Learn More
ACROBiosystems Human Transferrin Protein, premium grade
Catalog No. 502718808
Encompass_Preferred
Transferrin is also known as Serotransferrin, Beta-1 metal-binding globulin, TF, and is iron-binding blood plasma glycoproteins that control the level of free iron in biological fluids. Although iron bound to transferrin is less than 0.1% (4 mg) of the total body iron, it is the most important iron pool, with the highest rate of turnover (25 mg/24 h). The affinity of transferrin for Fe(III) is extremely high (1023 M−1 at pH 7.4) but decreases progressively with decreasing pH below neutrality.When not bound to iron, it is known as "apo-transferrin”. In humans, transferrin consists of a polypeptide chain containing 679 amino acids. It is a complex composed of alpha helices and beta sheets to form two domains (the first situated in the N-terminus and the second in the C-terminus). The N- and C- terminal sequences are represented by globular lobes and between the two lobes is an iron-binding site. The liver is the main source of manufacturing transferrin,
Catalog No. 50-271-8808 Supplier ACROBiosystems Supplier No. TRNH5216200UG
Transferrin is also known as Serotransferrin, Beta-1 metal-binding globulin, TF, and is iron-binding blood plasma glycoproteins that control the level of free iron in biological fluids. Although iron bound to transferrin is less than 0.1% (4 mg) of the total body iron, it is the most important iron pool, with the highest rate of turnover (25 mg/24 h). The affinity of transferrin for Fe(III) is extremely high (1023 M−1 at pH 7.4) but decreases progressively with decreasing pH below neutrality.When not bound to iron, it is known as "apo-transferrin”. In humans, transferrin consists of a polypeptide chain containing 679 amino acids. It is a complex composed of alpha helices and beta sheets to form two domains (the first situated in the N-terminus and the second in the C-terminus). The N- and C- terminal sequences are represented by globular lobes and between the two lobes is an iron-binding site. The liver is the main source of manufacturing transferrin,