Learn More
Rockland Immunochemicals HUMAN TRANSFERRIN - 009-0134
SDP
Supplier:  Rockland Immunochemicals 0090134
Encompass
Human transferrin is encoded by the TF gene and is an iron-binding blood plasma glycoprotein that controls the level of free iron in biological fluids. Human transferrin binds iron very tightly but reversibly. Human transferrin is the most important iron pool in mammals. Human transferrin has a molecular weight of around 80 kDa and contains 2 specific high-affinity Fe(III) binding sites. The affinity of Human transferrin for Fe(III) is extremely high but decreases progressively with decreasing pH below neutrality.
Catalog No. 50-105-7331