Learn More
ACROBiosystems Human Transferrin 25ug
Supplier:  ACROBiosystems TRNH82E325UG
Encompass_Preferred
Biotinylated Human Transferrin Protein HisAvitag (TRN-H82E3) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 698 (Accession # P02787).
Catalog No. 50-253-4008