Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human Transferrin R Protein, Tag Free, Lyophilized, 1 mg

Catalog No. 502718835
Encompass_Preferred

Human Transferrin receptor protein 1 (CD71) is a critical protein required for iron delivery from transferrin to cells. Belonging to the peptidase M28 family, this protein is expressed in human HEK293 cells and contains one protease-associated (PA) domain. It is recognized as a potential target in human leukemia and lymphoma research and has been shown to interact with GABARAP and HFE.

  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS
  • Molecular weight of 75.2 kDa as calculated, migrating at 90-95 kDa due to glycosylation
  • Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose
  • Contains amino acids Cys 89 - Phe 760
  • Endotoxin level of less than 0.01 EU per μg
  • Designed as a dimer
  • Validated applications include ELISA and SPR

Catalog No. 50-271-8835 Supplier ACROBiosystems Supplier No. TFRH52131MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human Transferrin receptor protein 1 (CD71) is a critical protein required for iron delivery from transferrin to cells. Belonging to the peptidase M28 family, this protein is expressed in human HEK293 cells and contains one protease-associated (PA) domain. It is recognized as a potential target in human leukemia and lymphoma research and has been shown to interact with GABARAP and HFE.

  • Greater than 95% purity as determined by SDS-PAGE and SEC-MALS
  • Molecular weight of 75.2 kDa as calculated, migrating at 90-95 kDa due to glycosylation
  • Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose
  • Contains amino acids Cys 89 - Phe 760
  • Endotoxin level of less than 0.01 EU per μg
  • Designed as a dimer
  • Validated applications include ELISA and SPR

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.