Learn More
ACROBiosystems Human Transferrin R / CD71 Protein, Fc Tag
Catalog No. 502331367
Encompass_Preferred
Human Transferrin R Protein, Fc Tag (TFR-H5264) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1).
Catalog No. 50-233-1367 Supplier ACROBiosystems Supplier No. TFRH52641MG
Human Transferrin R Protein, Fc Tag (TFR-H5264) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1).