Learn More
ACROBiosystems Human Transferrin R 200ug
Catalog No. 502534066
Encompass_Preferred
Biotinylated Human Transferrin R Protein FcAvitag (TFR-H82F3) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1).
Catalog No. 50-253-4066 Supplier ACROBiosystems Supplier No. TFRH82F3200UG
Biotinylated Human Transferrin R Protein FcAvitag (TFR-H82F3) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession # P02786-1).