Learn More
American Research Products Inc Human Transforming Growth fact
Catalog No. 5000329179
Encompass
Human Transforming Growth factor b1 TGF-b1 ELISA kit from CUSABIO Detection Range 23 5 pg/ml-1500 pg/ml Sensitivity 5 8 pg/ml Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-29179 Supplier American Research Products Inc Supplier No. CSBE04725HIS
Human Transforming Growth factor b1 TGF-b1 ELISA kit from CUSABIO Detection Range 23 5 pg/ml-1500 pg/ml Sensitivity 5 8 pg/ml Sample serum plasma cell culture supernates Method Sandwich