Learn More
Biomatik Corporation Human Transforming Growth Factor Beta 1 (TGFb1) CLIA Kit
Supplier:  Biomatik Corporation EKU08838
Encompass_Preferred
Human Transforming Growth Factor Beta 1 (TGFb1) CLIA Kit for detecting TGFb1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.96pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01137. Gene ID: 7040. Target Synonyms: TGF-B1; CED; DPD1; LAP; Camurati-Engelmann Disease; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6124