Learn More
Biomatik Corporation Human Transforming Growth Factor Beta Stimulated Protein Clone 22 (TSC22) ELISA Kit
Supplier:  Biomatik Corporation EKU07825
Encompass_Preferred
Human Transforming Growth Factor Beta Stimulated Protein Clone 22 (TSC22) ELISA Kit for detecting TSC22 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15714. Gene ID: 8848. Target Synonyms: TSC22D1; TGFB1I4; TSC22; hucep-2; Cerebral protein 2; Transforming Growth Factor Beta 1 Induced Transcript 4; TSC22 Domain Family,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6650