Learn More
Biomatik Corporation Human Transforming Growth Factor Alpha (TGFa) CLIA Kit
Supplier:  Biomatik Corporation EKU08837
Encompass_Preferred
Human Transforming Growth Factor Alpha (TGFa) CLIA Kit for detecting TGFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.51pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01135. Gene ID: 7039. Target Synonyms: ETGF; TGF-A; Protransforming Growth Factor alpha; EGF-Like TGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8615