Learn More
Biomatik Corporation Human Transforming Growth Factor Β1, Tgf-Β1 Elisa Kit
Supplier:  Biomatik Corporation EKC31341
Encompass_Preferred
Human Transforming Growth Factor Β1, Tgf-Β1 Elisa Kit for detecting Transforming Growth Factor, Beta 1 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.747ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01137; TGFB1. Target Synonyms: Cartilage-inducing factor; CED; Differentiation inhibiting factor; DPD1; LAP; Latency-associated peptide; Prepro transforming growth factor beta 1; TGF beta 1; TGF beta; TGF beta 1 protein; TGF-beta 1 protein; TGF-beta-1; TGF-beta-5; TGF-beta1; TGFB; Tgfb-1; tgfb1; TGFB1_HUMAN; TGFbeta; TGFbeta1; Transforming Growth Factor b1; Transforming Growth Factor beta 1; Transforming growth factor beta 1a; transforming growth factor beta-1; transforming growth factor; beta 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9192