Learn More
Cloud Clone Corp Human Transforming Growth Factor Beta 1 (TGFb1) (Sandwich ELISA) ELISA Kit
Catalog No. NC4011152
Encompass_Preferred
Human Transforming Growth Factor Beta 1 (TGFb1) (Sandwich ELISA) ELISA Kit measures Human TGFb1 in Serum, Platelet-Poor Plasma, Tissue homogenates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Transforming Growth Factor Beta 1 (TGFb1).No significant cross-reactivity or interference between Transforming Growth Factor Beta 1 (TGFb1) and analogues was observed.Detection Range: 15.6-1,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 5.7pg/mLDetection menthod: ColormetricGene ID:7040
Catalog No. NC4011152 Supplier Cloud Clone Corp Supplier No. SEA124HU
Human Transforming Growth Factor Beta 1 (TGFb1) (Sandwich ELISA) ELISA Kit measures Human TGFb1 in Serum, Platelet-Poor Plasma, Tissue homogenates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Transforming Growth Factor Beta 1 (TGFb1).No significant cross-reactivity or interference between Transforming Growth Factor Beta 1 (TGFb1) and analogues was observed.Detection Range: 15.6-1,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 5.7pg/mLDetection menthod: ColormetricGene ID:7040