Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Transglutaminase 3/TGM3 ELISA Kit

Catalog No. EH463RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH463RB 96 Tests
EH463RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH463RB Supplier Invitrogen™ Supplier No. EH463RB
Add to Cart
Add to Cart

ELISA

Human Transglutaminase 3/TGM3 quantitates human Transglutaminase 3/TGM3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Transglutaminase 3/TGM3.

Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene consists of two polypeptide chains activated from a single precursor protein by proteolysis. The encoded protein is involved the later stages of cell envelope formation in the epidermis and hair follicle.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q08188
Assay Range 0.16-40 ng/mL
Assay Sensitivity 0.16 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI893889; E polypeptide; E polypeptide, protein-glutamine-gamma-glutamyltransferase; MGC126249; MGC126250; protein-glutamine gamma-glutamyltransferase E; Protein-glutamine gamma-glutamyltransferase E 27 kDa non-catalytic chain; Protein-glutamine gamma-glutamyltransferase E 50 kDa catalytic chain; protein-glutamine-gamma-glutamyltransferase; RGD1561831; TG E; TG(E); TGase E; TGase TGase-3; Tgase3; TGase-3; TGE; TGM3; transglutaminase 3; transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase); transglutaminase 3, E polypeptide; transglutaminase E; Transglutaminase-3AI893889; E polypeptide; E polypeptide, protein-glutamine-gamma-glutamyltransferase; MGC126249; MGC126250; protein-glutamine gamma-glutamyltransferase E; Protein-glutamine gamma-glutamyltransferase E 27 kDa non-catalytic chain; Protein-glutamine gamma-glutamyltransferase E 50 kDa catalytic chain; protein-glutamine-gamma-glutamyltransferase; RGD1561831; TG E; TG(E); TGase E; TGase TGase-3; Tgase3; TGase-3; TGE; TGM3; transglutaminase 3; transglutaminase 3 (E polypeptide, protein-glutamine-gamma-glutamyltransferase); transglutaminase 3, E polypeptide; transglutaminase E; Transglutaminase-3;
Gene ID (Entrez) 7053
Gene Symbol TGM3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target TGM3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.