Learn More
Biomatik Corporation Human Transient Receptor Potential Cation Channel Subfamily V, Member 3 (TRPV3) ELISA Kit
Supplier:  Biomatik Corporation EKU07847
Encompass_Preferred
Human Transient Receptor Potential Cation Channel Subfamily V, Member 3 (TRPV3) ELISA Kit for detecting TRPV3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8NET8. Gene ID: 162514. Target Synonyms: VRL3; Vanilloid receptor-like 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4941