Learn More
Biomatik Corporation Human Transketolase Like Protein 1 (TKTL1) ELISA Kit
Supplier:  Biomatik Corporation EKU07850
Encompass_Preferred
Human Transketolase Like Protein 1 (TKTL1) ELISA Kit for detecting TKTL1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51854. Gene ID: 8277. Target Synonyms: TKR; TKT2; TK 2; Transketolase-related protein; Transketolase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6812