Learn More
Biomatik Corporation Human Transmembrane Protein 173 (TMEM173) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU12094
Encompass_Preferred
Human Transmembrane Protein 173 (TMEM173) High-sensitive ELISA Kit for detecting TMEM173 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86WV6. Gene ID: 340061. Target Synonyms: NET23; ERIS; MPYS; MITA; STING; Endoplasmic Reticulum Interferon Stimulator; Stimulator Of Interferon Genes Protein; Mediator of IRF3 activation. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6280