Learn More
Biomatik Corporation Human Transportin 2 (TNPO2) ELISA Kit
Supplier:  Biomatik Corporation EKU07863
Encompass_Preferred
Human Transportin 2 (TNPO2) ELISA Kit for detecting TNPO2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O14787. Gene ID: 30000. Target Synonyms: IPO3; KPNB2B; TRN2; Importin 3; Karyopherin Beta 2b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8718