Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human TRBC1 Protein, His, Avitag, 25 μg

Catalog No. 502370266
Encompass_Preferred

Biotinylated Human TRBC1 Protein (His, Avitag) is a recombinant fragment of the human T cell receptor beta constant 1 (Asp 1-Ala 144, Accession P01850-1) expressed in HEK293 cells and enzymatically biotinylated for use in T-cell receptor research and assay development. Supplied lyophilized in 25 μg vials.

  • Recombinant fragment comprising Asp 1-Ala 144 (Accession P01850-1).
  • Expressed in HEK293 (human embryonic kidney) cells.
  • C-terminal polyhistidine tag with enzymatic biotinylation at a biotin acceptor peptide.
  • Calculated molecular weight 19.9 kDa; migrates at ~23 kDa and ~26 kDa
  • Purity greater than 90%
  • Endotoxin less than 0.2 EU/μg (LAL / rFC).
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Store lyophilized at -20°C or lower; 12-month stability lyophilized at -20°C to -70°C; reconstituted stability up to 3 months at -70°C under sterile conditions.

Catalog No. 50-237-0266 Supplier ACROBiosystems Supplier No. TR1H82E625UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated Human TRBC1 Protein (His, Avitag) is a recombinant fragment of the human T cell receptor beta constant 1 (Asp 1-Ala 144, Accession P01850-1) expressed in HEK293 cells and enzymatically biotinylated for use in T-cell receptor research and assay development. Supplied lyophilized in 25 μg vials.

  • Recombinant fragment comprising Asp 1-Ala 144 (Accession P01850-1).
  • Expressed in HEK293 (human embryonic kidney) cells.
  • C-terminal polyhistidine tag with enzymatic biotinylation at a biotin acceptor peptide.
  • Calculated molecular weight 19.9 kDa; migrates at ~23 kDa and ~26 kDa
  • Purity greater than 90%
  • Endotoxin less than 0.2 EU/μg (LAL / rFC).
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Store lyophilized at -20°C or lower; 12-month stability lyophilized at -20°C to -70°C; reconstituted stability up to 3 months at -70°C under sterile conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.