Learn More
ACROBiosystems Human TRBC1, His Tag (TR1-H52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 1 - Ala 144 (Accession # P01850-1).
Supplier:  ACROBiosystems TR1H52H4100UG
Encompass_Preferred
The transmembrane protein, TCR, comprise of two disulphide-linked polypeptide chains: a α and β chain, a γ and δ chain. Each polypeptide chain consists of a variable and a constant region. TRBC1 is the constant region of T-cell receptor (TCR) beta chain. TRBC1 is presented on the surface of T cell and recognized peptide-major histocompatibility (MH) (pMH) that are displayed by antigen presenting cells (APC). TRBC1 is participate in an adaptive immune response and has been well-studied in T cell therapy.
Catalog No. 50-225-4047