Learn More
Biomatik Corporation Human Trefoil Factor 3 (TFF3) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10965
Encompass_Preferred
Human Trefoil Factor 3 (TFF3) High-sensitive ELISA Kit for detecting TFF3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 14.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07654. Gene ID: 7033. Target Synonyms: HITF; ITF3; TFI; HP1.B; Trefoil Factor 3, Intestinal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8261