Learn More
Sino Biological Anti-TREM-1 Antibody (Biotin), Mouse Monoclonal
Supplier:  Sino Biological 10511MM07B100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TREM-1 / TREM1 (rh TREM-1 / TREM1; Catalog#10511-H08H; NP_061113.1; Met1-Arg200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-241-2272