Learn More
Sino Biological Recombinant Human TREM-1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10511H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg 200) of human TREM1 (NP_061113.1) was fused with the a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5273