Learn More
Abcam Human TREM2 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2248811X96TESTS
Encompass_Preferred
Human TREM2 ELISA Kit is a single-wash 90-min SimpleStep Sandwich ELISA used to quantify Human TREM2 with a sensitivity of 10.5 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout, works on any plate reader - Validated on a number of sample types including cerebrospinal fluid (CSF) - Need to design your own assay: we also offer the conjugation-ready antibody pair - Cited in over 10 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2384805