Learn More
Sino Biological Recombinant Anti-TREM-2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11084R016100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human TREM-2 / TREM2 (rh TREM-2 / TREM2; Catalog#11084-H08H; NP_061838.1; Met1-Ser174).
Catalog No. 50-241-1281