Learn More
Sino Biological Anti-TREM-2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11084MM0820
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TREM-2 / TREM2 (rh TREM-2 / TREM2; Catalog#11084-H08H; NP_061838.1; Met1-Ser174). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8606