Learn More
Sino Biological Recombinant Human TREM-2 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 11084H02H100
Encompass_Preferred
A DNA sequence encoding the human TREM2 (NP_061838.1) (Met1-Ser174) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2228