Learn More
ACROBiosystems Human TREM2 Protein, Mouse IgG2a Fc Tag
Catalog No. 502054906
Encompass_Preferred
Recombinant Protein;1MG;Human TREM2, Mouse IgG2a Fc Tag (TR2-H5256) is expressed from human 293 cells (HEK293). It contains AA His 19 - Ser 174 (Accession # Q9NZC2-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TREM2, Mouse IgG2a Fc Tag (Cat. No. TR2-H5256) at 1 μg/mL (100 μL/well) can bind Anti-TREM2 Antibody, Human IgG1 with a linear range of 0.2-5 ng/mL (QC tested).
Catalog No. 50-205-4906 Supplier ACROBiosystems Supplier No. TR2H52561MG
Recombinant Protein;1MG;Human TREM2, Mouse IgG2a Fc Tag (TR2-H5256) is expressed from human 293 cells (HEK293). It contains AA His 19 - Ser 174 (Accession # Q9NZC2-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TREM2, Mouse IgG2a Fc Tag (Cat. No. TR2-H5256) at 1 μg/mL (100 μL/well) can bind Anti-TREM2 Antibody, Human IgG1 with a linear range of 0.2-5 ng/mL (QC tested).