Learn More
Sino Biological Anti-TLT-1/TREML1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 204035T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human TLT-1/TREML1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2035