Learn More
Sino Biological Anti-TLT-2/TREML2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11655RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human TREML2 (rh TREML2; Catalog#11655-H08H; NP_079083.2; Met 1-Ser 268). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4644