Learn More
American Research Products Inc Human Triggering receptor eXpr
Catalog No. 5000327586
Encompass
Human Triggering receptor eXpressed on myeloid cells 2 (TREM2) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27586 Supplier American Research Products Inc Supplier No. CSBEL024405HU
Add to Cart
Add to Cart
Human Triggering receptor eXpressed on myeloid cells 2 (TREM2) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich