Learn More
Biomatik Corporation Human Triggering Receptor Expressed On Myeloid Cells 1 (TREM1) CLIA Kit
Supplier:  Biomatik Corporation EKU08859
Encompass_Preferred
Human Triggering Receptor Expressed On Myeloid Cells 1 (TREM1) CLIA Kit for detecting TREM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9NP99. Gene ID: 54210. Target Synonyms: CD354; Triggering Receptor Expressed On Monocytes-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6795