Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human TRIM67 (aa 9-85) Control Fragment Recombinant Protein

Catalog No. RP94143
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP94143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP94143 Supplier Invitrogen™ Supplier No. RP94143
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57056. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The TRIM67 gene encodes a member of the tripartite motif (TRIM) protein family, which is involved in various cellular processes including protein ubiquitination, signal transduction, and immune responses. TRIM67 contains RING, B-box, and coiled-coil domains, characteristic of TRIM proteins, which facilitate interactions with other proteins and contribute to its role in ubiquitination and cellular regulation. TRIM67 is notably expressed in the brain and has been associated with neurodevelopmental processes, particularly influencing neuronal differentiation and synapse formation. This gene plays a regulatory role in signaling pathways that are critical for neural development, and mutations or dysregulation in TRIM67 have been linked to neurodevelopmental disorders and cognitive deficits. Additionally, TRIM67's involvement in protein degradation and signal modulation may impact cellular architecture and brain function. Ongoing research seeks to elucidate TRIM67's precise mechanisms and potential therapeutic targets for neurological conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6ZTA4
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 440730
Name Human TRIM67 (aa 9-85) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias D130049O21Rik; RGD1560667; TNL; Trim67; TRIM9-like protein; TRIM9-like protein TNL; tripartite motif containing 67; tripartite motif-containing 67; tripartite motif-containing protein 67
Common Name TRIM67
Gene Symbol TRIM67
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VCGSLFREPIILPCSHNVCLPCARTIAVQTPDGEQHLPQPLLLSRGSGLQAGAAAAASLEHDAAAGPACGGAGGSAA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.